accessibility menu, dialog, popup

UserName

H3CONHNH2O

CAS RN 2620-63-5

IUPAC Name: 2-acetamidoacetamide
Synonyms: N-acetylglycinamide
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: WQELDIQOHGAHEM-UHFFFAOYSA-N
SMILES: CC(=O)NCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results