Learn More
N-Acetylglycinamide, 97%
Product Code. 11491457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491457 5 g 5g
11481457 1 g 1g
11401467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11491457 Supplier Thermo Scientific Alfa Aesar Supplier No. B24940.06
Request Bulk or Custom Format
CAS: 2620-63-5 | C4H8N2O2 | 116.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2620-63-5
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008029
InChI Key WQELDIQOHGAHEM-UHFFFAOYSA-N
Synonym n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide
PubChem CID 28326
IUPAC Name 2-acetamidoacetamide
SMILES CC(=O)NCC(N)=O
Specifications
Melting Point 136°C to 140°C
Odor Odorless
Quantity 5 g
Beilstein 1753982
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material N-Acetylglycinamide