CAS RN 26306-06-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deoxybenzoin Oxime, TRC
CAS: 26306-06-9 Molecular Formula: C14 H13 N O Molecular Weight (g/mol): 211.26 Synonym: N-(1,2-Diphenylethylidene)hydroxylamine,(1Z)-1,2-Diphenyl-ethanone Oxime IUPAC Name: (NZ)-N-(1,2-diphenylethylidene)hydroxylamine SMILES: O\N=C(\Cc1ccccc1)/c2ccccc2