missing translation for 'onlineSavingsMsg'
Learn More

Deoxybenzoin Oxime, TRC

Product Code. 30365044
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.50g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365044 2.5 g 2.50g
30356190 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365044 Supplier Toronto Research Chemicals Supplier No. TRCD2322502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26306-06-9
Molecular Formula C14 H13 N O
Molecular Weight (g/mol) 211.26
Synonym N-(1,2-Diphenylethylidene)hydroxylamine, (1Z)-1,2-Diphenyl-ethanone Oxime
IUPAC Name (NZ)-N-(1,2-diphenylethylidene)hydroxylamine
SMILES O\N=C(\Cc1ccccc1)/c2ccccc2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H13NO/c16-15-14(13-9-5-2-6-10-13)11-12-7-3-1-4-8-12/h1-10,16H,11H2/b15-14-
Formula Weight 211.0997
Chemical Name or Material Deoxybenzoin Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.