accessibility menu, dialog, popup

UserName

H3CCH3HN(S)OHONH

CAS RN 26328-11-0

IUPAC Name: (2S)-1-(1H-indol-4-yloxy)-3-[(propan-2-yl)amino]propan-2-ol
Synonyms: (-)-pindolol (S)-pindolol (S)-(-)-pindolol
Molecular Weight (g/mol): 248.33
Molecular Formula: C14H20N2O2
InChi Key: JZQKKSLKJUAGIC-NSHDSACASA-N
SMILES: CC(C)NC[C@H](O)COC1=C2C=CNC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results