missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Pindolol, Tocris Bioscience™

Product Code. 30001386
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001386 10 mg 10mg
18075752 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001386 Supplier Tocris Bioscience Supplier No. 1060/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

β3 partial agonist; also 5-HT1A and 5-HT1B antagonist; more active enantiomer of pindolol (Cat. No. 0994)

5-HT1A/1B receptor antagonist, with roughly equal affinity for each subtype. A partial agonist at mouse and human β3-adrenoceptors. More active enantiomer of pindolol .

Racemate also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26328-11-0
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.326
InChI Key JZQKKSLKJUAGIC-NSHDSACASA-N
Synonym s--pindolol, s---pindolol, --pindolol, s-pindolol, s ?-pindolol, 2s-1-1h-indol-4-yloxy-3-isopropylamino propan-2-ol, 2s-1-1h-indol-4-yloxy-3-1-methylethyl amino propan-2-ol, 2s-1-1h-indol-4-yloxy-3-propan-2-yl amino propan-2-ol, 2s-1-1h-indol-4-yloxy-3-propan-2-ylamino propan-2-ol, s-1-1h-indol-4-yloxy-3-1-methylethyl amino-2-propanol
PubChem CID 688095
ChEBI CHEBI:48281
IUPAC Name (2S)-1-(1H-indol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(COC1=CC=CC2=C1C=CN2)O

Specifications

Quantity 10 mg
Formula Weight 248.32
Percent Purity >99%
Chemical Name or Material (S)-(-)-Pindolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.