accessibility menu, dialog, popup

UserName

MgBrH3CCH3CH3

CAS RN 2633-66-1

IUPAC Name: magnesium(2+) 2,4,6-trimethylbenzen-1-ide bromide
Synonyms: magnesium(2+) 2,4,6-trimethylbenzen-1-ide bromide
Molecular Weight (g/mol): 223.4
Molecular Formula: C9H11BrMg
InChi Key: JOWQNXIISCPKBK-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=CC(C)=[C-]C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results