accessibility menu, dialog, popup

UserName

H3CNNOOFFFNOO

CAS RN 26399-36-0

IUPAC Name: N-(cyclopropylmethyl)-2,6-dinitro-N-propyl-4-(trifluoromethyl)aniline
Synonyms: profluralin
Molecular Weight (g/mol): 347.29
Molecular Formula: C14H16F3N3O4
InChi Key: ITVQAKZNYJEWKS-UHFFFAOYSA-N
SMILES: CCCN(CC1CC1)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Concentration 
  • (1)
  • (1)
Element 
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results