CAS RN 2641-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Phosphoglucono-Delta-lactone, TRC
CAS: 2641-81-8 Molecular Formula: C6H11O9P . xNH 3 Molecular Weight (g/mol): 258.12 Synonym: D-Gluconic Acid δ-Lactone 6-(Dihydrogen Phosphate) Ammonium Salt,6-Phospho-D-glucono-1,5-lactone,6-Phosphoglucono-δ-lactone Ammonium Salt,δ-Gluconolactone 6-Phosphate Ammonium Salt SMILES: O[C@@H]1[C@H]([C@@H]([C@H](OC1=O)COP(O)(O)=O)O)O.O=C2[C@@H]([C@H]([C@H](O2)[C@@H](COP(O)(O)=O)O)O)O.N.N