Learn More
6-Phosphoglucono-Delta-lactone, TRC
Product Code. 30420711
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420711 1 mg -
30420341 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30420711 Supplier Toronto Research Chemicals Supplier No. TRCP3537401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2641-81-8
Molecular Formula C6H11O9P . xNH 3
Molecular Weight (g/mol) 258.12
Synonym D-Gluconic Acid δ-Lactone 6-(Dihydrogen Phosphate) Ammonium Salt, 6-Phospho-D-glucono-1,5-lactone, 6-Phosphoglucono-δ-lactone Ammonium Salt, δ-Gluconolactone 6-Phosphate Ammonium Salt
SMILES O[C@@H]1[C@H]([C@@H]([C@H](OC1=O)COP(O)(O)=O)O)O.O=C2[C@@H]([C@H]([C@H](O2)[C@@H](COP(O)(O)=O)O)O)O.N.N
Specifications
Quantity 1 mg
InChI Formula InChI=1S/2C6H11O9P.2H3N/c7-2(1-14-16(11,12)13)5-3(8)4(9)6(10)15-5;7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;/h2*2-5,7-9H,1H2,(H2,11,12,13);2*1H3/t2-,3-,4-,5-;2-,3-,4+,5-;;/m11../s1
Percent Purity >95%
Chemical Name or Material 6-Phosphoglucono-delta-lactone
Grade HPLC