CAS RN 26411-50-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycidyl Caprate, TRC
CAS: 26411-50-7 Molecular Formula: C13H24O3 Molecular Weight (g/mol): 228.33 Synonym: Decanoic Acid 2-Oxiranylmethyl Ester,Decanoic Acid 2,3-Epoxypropyl Ester,2,3-Epoxy-1-propanol Decanoate,Glycidyl Decanoate; IUPAC Name: oxiran-2-ylmethyl decanoate SMILES: CCCCCCCCCC(=O)OCC1CO1