accessibility menu, dialog, popup

UserName

H3COOON

CAS RN 26510-52-1

IUPAC Name: ethyl 3-oxo-3-(pyridin-2-yl)propanoate
Synonyms: ethyl 3-oxo-3-(pyridin-2-yl)propanoate
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: FQHXWZMJALFSJJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=CC=N1
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results