CAS RN 26537-19-9
IUPAC Name:
methyl 4-tert-butylbenzoate
Synonyms:
methyl 4-T-butylbenzoate
Molecular Weight (g/mol):
192.26
Molecular Formula:
C12H16O2
InChi Key:
UPIJOAFHOIWPLT-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1)C(C)(C)C
1
–
2
of
2
results
2
Methyl 4-tert-butylbenzoate, 98+%
CAS: 26537-19-9 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.258 MDL Number: MFCD00008835 InChI Key: UPIJOAFHOIWPLT-UHFFFAOYSA-N Synonym: methyl 4-tert-butyl benzoate,methyl p-tert-butylbenzoate,4-tert-butylbenzoic acid methyl ester,unii-8vk4o27jmt,benzoic acid, 4-1,1-dimethylethyl-, methyl ester,methyl 4-t-butylbenzoate,8vk4o27jmt,methyl4-tert-butylbenzoate,methyl p-t-butylbenzoate,benzoic acid, p-tert-butyl-, methyl ester PubChem CID: 97433 IUPAC Name: methyl 4-tert-butylbenzoate SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)OC