CAS RN 26550-55-0
IUPAC Name:
1-methoxypropan-2-ol
Synonyms:
methoxyisopropanol
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
ARXJGSRGQADJSQ-UHFFFAOYNA-N
SMILES:
COCC(C)O
1
–
1
of
1
results
1
(S)-(+)-1-Methoxy-2-propanol, 99%
CAS: 26550-55-0 Molecular Formula: C4H10O2 Molecular Weight (g/mol): 90.12 MDL Number: MFCD01632588 InChI Key: ARXJGSRGQADJSQ-BYPYZUCNSA-N Synonym: s-+-1-methoxy-2-propanol,2s-1-methoxypropan-2-ol,s-1-methoxypropan-2-ol,s---1-methoxy-2-propanol,s-1-methoxy-2-propanol,2-propanol, 1-methoxy-, 2s,2s-1-methoxy-2-propanol,pubchem6715,s-1-methoxy-propan-2-ol PubChem CID: 6993459 IUPAC Name: (2S)-1-methoxypropan-2-ol SMILES: CC(COC)O