CAS RN 26586-55-0
IUPAC Name:
1-[4-(4-methylpiperazin-1-yl)phenyl]ethan-1-one
Synonyms:
1-[4-(4-methylpiperazin-1-yl)phenyl]ethanone
Molecular Weight (g/mol):
218.3
Molecular Formula:
C13H18N2O
InChi Key:
IRIZGAMYKHTLKS-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C1=CC=C(C=C1)C(C)=O