accessibility menu, dialog, popup

UserName

CH3NHOOOHO

CAS RN 26607-51-2

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: TYRGLVWXHJRKMT-UHFFFAOYNA-N
SMILES: CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)