Learn More
N-Benzyloxycarbonyl-D-alanine, 98+%, Thermo Scientific Chemicals
Product Code. 11368125
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368125 5 g 5g
11358125 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11368125 Supplier Thermo Scientific Alfa Aesar Supplier No. L08382.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26607-51-2
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00063126
InChI Key TYRGLVWXHJRKMT-UHFFFAOYNA-N
Synonym n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
PubChem CID 70832
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Specifications
Melting Point 83°C to 86°C
Quantity 5 g
Beilstein 2056163
Optical Rotation +14.5° (c=2 in Acetic Acid)
Formula Weight 223.23
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-alanine