accessibility menu, dialog, popup

UserName

H3CCH3H3COOOOCH3NH2HCl

CAS RN 2673-19-0

IUPAC Name: 4-tert-butyl 1-methyl 2-aminobutanedioate hydrochloride
Synonyms: 4-tert-butyl 1-methyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol): 239.7
Molecular Formula: C9H18ClNO4
InChi Key: SFYKWYAIJZEDNG-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)