accessibility menu, dialog, popup

UserName

CH3OH2COHONHOH

CAS RN 26750-81-2

IUPAC Name: 2-hydroxy-N-(2-hydroxyethyl)-3-methoxy-5-(prop-2-en-1-yl)benzamide
Synonyms: 2-hydroxy-N-(2-hydroxyethyl)-3-methoxy-5-(prop-2-en-1-yl)benzamide
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: UMJHTFHIQDEGKB-UHFFFAOYSA-N
SMILES: COC1=CC(CC=C)=CC(C(=O)NCCO)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)