CAS RN 26761-40-0
IUPAC Name:
1,2-bis(8-methylnonyl) benzene-1,2-dicarboxylate
Synonyms:
diisodecyl phthalate Didp palatinol Z didp (plasticizer) sicol 184 bis(isodecyl)phthalate plasticized ddp vestinol DZ di-''isodecyl'' phthalate
Molecular Weight (g/mol):
446.67
Molecular Formula:
C28H46O4
InChi Key:
ZVFDTKUVRCTHQE-UHFFFAOYSA-N
SMILES:
CC(C)CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC(C)C
1
–
4
of
4
results
1
Diisodecyl Phthalate (Mixture of Branched chain Isomers) (>80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diisodecyl Phthalate (Mixture of Branched chain Isomers) (>80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diisodecyl Phthalate (Mixture of Branched chain Isomers) (>80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.