accessibility menu, dialog, popup

UserName

NH3(R)OO

CAS RN 26774-88-9

IUPAC Name: (2R)-2-azaniumyl-2-(cyclohexa-1,4-dien-1-yl)acetate
Synonyms: (2R)-2-ammonio-2-(cyclohexa-1,4-dien-1-yl)acetate
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: JBJJTCGQCRGNOL-SSDOTTSWSA-N
SMILES: [NH3+][C@@H](C([O-])=O)C1=CCC=CC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)