Learn More
(-)-2,5-Dihydro-D-phenylglycine, 97%, Thermo Scientific™
Product Code. 11353247
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353247 100 g 100g
11333247 5 g 5g
11343247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11353247 Supplier Thermo Scientific Alfa Aesar Supplier No. L13785.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 26774-88-9
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00137746
InChI Key JBJJTCGQCRGNOL-SSDOTTSWSA-N
Synonym r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine
PubChem CID 1268102
IUPAC Name (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid
SMILES C1C=CCC(=C1)C(C(=O)O)N
Specifica
Melting Point 288°C to 290°C (sublimation)
Beilstein 2091420
Quantity 100 g
Formula Weight 153.18
Optical Rotation −160° (c=1 in 1N HCl)
Percent Purity 97%
Chemical Name or Material (-)-2,5-Dihydro-D-phenylglycine
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.