CAS RN 2679-87-0
IUPAC Name:
2-ethoxybutane
Synonyms:
butane, 2-ethoxy-
Molecular Weight (g/mol):
102.18
Molecular Formula:
C6H14O
InChi Key:
VSCUCHUDCLERMY-UHFFFAOYNA-N
SMILES:
CCOC(C)CC
1
–
1
of
1
results
1
ASTM Method D7796 Impurities in ETBE 0.1 - 93 Wt %, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality