CAS RN 268550-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.