CAS RN 2688-48-4
IUPAC Name:
5-hydroxy-2,3-dihydro-1-benzofuran-2-one
Synonyms:
5-hydroxy-3H-1-benzofuran-2-one
Molecular Weight (g/mol):
150.13
Molecular Formula:
C8H6O3
InChi Key:
POUITAHNNRJWMA-UHFFFAOYSA-N
SMILES:
OC1=CC=C2OC(=O)CC2=C1
1
–
3
of
3
results
1
Homogentisic Acid gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Homogentisic Acid gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Homogentisic Acid gamma-Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.