CAS RN 27013-91-8
CAS RN 27013-91-8
IUPAC Name:
(4aS,6aS,6bR,8aR,9R,10S,12aR,12bR,14bS)-10-{[(2S,3R,4S,5S)-4,5-dihydroxy-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Synonyms:
α-hederine α-hederin helixin kalopanaxsaponin A sapindoside A tauroside E nepalin 2 α-hederin hydrate hederoside C koronaroside A
Molecular Weight (g/mol):
750.97
Molecular Formula:
C41H66O12
InChi Key:
KEOITPILCOILGM-LLJOFIFVSA-N
SMILES:
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)CO[C@H]2O[C@H]2CC[C@@]3(C)[C@@H](CC[C@]4(C)[C@@H]3CC=C3[C@@H]5CC(C)(C)CC[C@@]5(CC[C@@]43C)C(O)=O)[C@]2(C)CO)[C@H](O)[C@H](O)[C@H]1O
1
–
1
of
1
results
1
alpha-Hederin, MedChemExpress
MedChemExpress alpha-Hederin (α-Hederin), a monodesmosidic triterpenoid saponin, exhibits promising antitumor potential against a variety of human cancer cell lines. alpha-Hederin could inhibit the proliferation and induce apoptosis of gastric cancer accompanied by glutathione decrement and reactive oxygen species generation via activating mitochondrial dependent pathway.