accessibility menu, dialog, popup

UserName

CAS RN 2719-21-3

CH3ONHCH3O

CAS RN 2719-21-3

IUPAC Name: N-(4-acetylphenyl)acetamide
Synonyms: acetamide, N-(4-acetylphenyl)-
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: WECHHDJTILFYQT-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)