CAS RN 27208-80-6
CAS RN 27208-80-6
IUPAC Name:
(2S,3R,4S,5S,6R)-2-{3-hydroxy-5-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
piceid polydatin
Molecular Weight (g/mol):
390.39
Molecular Formula:
C20H22O8
InChi Key:
HSTZMXCBWJGKHG-CUYWLFDKSA-N
SMILES:
OC[C@H]1O[C@@H](OC2=CC(O)=CC(\C=C\C3=CC=C(O)C=C3)=C2)[C@H](O)[C@@H](O)[C@@H]1O
1
–
3
of
3
results
1
trans Resveratrol 3-O-beta-D-glucopyranoside, 98%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans Resveratrol 3-O-beta-D-glucopyranoside, 98%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Polydatin, MedChemExpress
MedChemExpress Polydatin (Piceid), extracted from the roots of Polygonum cuspidatum Sieb, a widely used traditional Chinese remedies, possesses anti-inflammatory activity in several experimental models. Polydatin (Piceid) inhibits G6PD and induces oxidative and ER stresses.