accessibility menu, dialog, popup

UserName

OOOOBr(E)

CAS RN 27452-00-2

IUPAC Name: (2E)-3-(6-bromo-2H-1,3-benzodioxol-5-yl)prop-2-enoate
Synonyms: (2E)-3-(6-bromo-2H-1,3-benzodioxol-5-yl)prop-2-enoate
Molecular Weight (g/mol): 270.06
Molecular Formula: C10H6BrO4
InChi Key: FIFYPFARGVBVGX-OWOJBTEDSA-M
SMILES: [O-]C(=O)\C=C\C1=CC2=C(OCO2)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results