Learn More
2-Bromo-4,5-methylenedioxycinnamic acid, 97%, Thermo Scientific™
Product Code. 11453737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453737 25 g 25g
11433737 1 g 1g
11443737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11453737 Supplier Thermo Scientific Alfa Aesar Supplier No. A18208.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27452-00-2
Molecular Formula C10H6BrO4
Molecular Weight (g/mol) 270.06
MDL Number MFCD00017599
InChI Key FIFYPFARGVBVGX-OWOJBTEDSA-M
Synonym 2-bromo-4,5-methylenedioxycinnamic acid, 2e-3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, 3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid, 6-bromo-3,4-methylenedioxy cinnamic acid, 2-propenoic acid,3-6-bromo-1,3-benzodioxol-5-yl, 3-6-bromo-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, bromobenzodioxolylacrylicacid, 2-bromo-4,5-methylenedioxy cinnamic acid, e-3-5-bromobenzo d 1,3 dioxol-6-yl acrylic acid, e-3-6-bromobenzo d 1,3 dioxol-5-yl acrylic acid
PubChem CID 6931592
IUPAC Name (E)-3-(6-bromo-1,3-benzodioxol-5-yl)prop-2-enoate
SMILES [O-]C(=O)\C=C\C1=CC2=C(OCO2)C=C1Br
Specifications
Melting Point ∼255°C (decomposition)
Quantity 25 g
Formula Weight 271.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-methylenedioxycinnamic acid