accessibility menu, dialog, popup

UserName

CAS RN 282116-97-6

BOOOCH3OHHO

CAS RN 282116-97-6

IUPAC Name: [4-(2-ethoxy-2-oxoethoxy)phenyl]boronic acid
Synonyms: 4-(2-ethoxy-2-oxoethoxy)phenylboronic acid
Molecular Weight (g/mol): 224.02
Molecular Formula: C10H13BO5
InChi Key: MECJJOCHJYCOJS-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)