accessibility menu, dialog, popup

UserName

CAS RN 289039-85-6

CH3OOICl

CAS RN 289039-85-6

IUPAC Name: methyl 3-chloro-5-iodobenzoate
Synonyms: methyl 3-chloro-5-iodobenzoate
Molecular Weight (g/mol): 296.49
Molecular Formula: C8H6ClIO2
InChi Key: ZDSIXLJPASGHDW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results