accessibility menu, dialog, popup

UserName

CAS RN 28917-82-0

CaOHOHOPOOO

CAS RN 28917-82-0

IUPAC Name: calcium 3-(phosphonatooxy)propane-1,2-diol
Synonyms: calcium glycerophosphate
Molecular Weight (g/mol): 210.14
Molecular Formula: C3H7CaO6P
InChi Key: IWIRHXNCFWGFJE-UHFFFAOYNA-L
SMILES: [Ca++].OCC(O)COP([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results