CAS RN 29218-07-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Deoxy-4-fluoro-D-glucose, TRC
CAS: 29218-07-3 Molecular Formula: C6 H11 F O5 Molecular Weight (g/mol): 182.15 Synonym: D-Glucose, 4-deoxy-4-fluoro-,4-Deoxy-4-fluoro-D-glucose,4-Deoxy-4-fluoro-D-glucopyranose IUPAC Name: (3R,4R,5S,6R)-5-fluoro-6-(hydroxymethyl)oxane-2,3,4-triol SMILES: OC[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1F