CAS RN 29427-48-3
CAS RN 29427-48-3
IUPAC Name:
1-(2-fluoro-4-methylphenyl)ethan-1-one
Synonyms:
1-(2-fluoro-4-methylphenyl)ethanone
Molecular Weight (g/mol):
152.17
Molecular Formula:
C9H9FO
InChi Key:
IBABHBXTLZNGAV-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(F)C=C(C)C=C1
1
–
1
of
1
results
1
2'-Fluoro-4'-methylacetophenone, 97%, Thermo Scientific™
CAS: 29427-48-3 Molecular Formula: C9H9FO Molecular Weight (g/mol): 152.17 MDL Number: MFCD03094331 InChI Key: IBABHBXTLZNGAV-UHFFFAOYSA-N Synonym: 1-2-fluoro-4-methylphenyl ethanone,2'-fluoro-4'-methylacetophenone,1-2-fluoro-4-methylphenyl ethan-1-one,ethanone, 1-2-fluoro-4-methylphenyl,acmc-1cjnp,4-acetyl-3-fluorotoluene,2-fluoro-4-methylacetophenone,1-acetyl-2-fluoro-4-methylbenzene,ethanone,1-2-fluoro-4-methylphenyl PubChem CID: 2778484 IUPAC Name: 1-(2-fluoro-4-methylphenyl)ethanone SMILES: CC(=O)C1=C(F)C=C(C)C=C1