accessibility menu, dialog, popup

UserName

CAS RN 29703-01-3

CsOOHO

CAS RN 29703-01-3

IUPAC Name: dicaesium(1+) carbonate
Synonyms: caesium carbonate carbonic acid, dicesium salt
Molecular Weight (g/mol): 325.82
Molecular Formula: CCs2O3
InChi Key: FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES: [Cs+].[Cs+].[O-]C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)