accessibility menu, dialog, popup

UserName

OHBrOBr

CAS RN 2973-77-5

IUPAC Name: 3,5-dibromo-4-hydroxybenzaldehyde
Synonyms: 3,5-dibromo-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: SXRHGLQCOLNZPT-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=O)C=C1Br
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)