accessibility menu, dialog, popup

UserName

CAS RN 30282-14-5

IUPAC Name: 2-hydroxycyclohexan-1-one
Synonyms: 2-hydroxycyclohexanone adipoin
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: ODZTXUXIYGJLMC-UHFFFAOYNA-N
SMILES: OC1CCCCC1=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)