CAS RN 30361-29-6
IUPAC Name:
(2E,4E)-undeca-2,4-dienal
Synonyms:
undecadienal
Molecular Weight (g/mol):
166.26
Molecular Formula:
C11H18O
InChi Key:
UVIUIIFPIWRILL-XBLVEGMJSA-N
SMILES:
CCCCCC\C=C\C=C\C=O
1
–
1
of
1
results
1
trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer
CAS: 30361-29-6 Molecular Formula: C11H18O Molecular Weight (g/mol): 166.264 MDL Number: MFCD00014677 InChI Key: UVIUIIFPIWRILL-XBLVEGMJSA-N Synonym: 2,4-undecadienal,2e,4e-undeca-2,4-dienal,undeca-2,4-dien-1-al,trans,trans-2,4-undecadien-1-al,2,4-undecadienal, 2e,4e,2,4-trans,trans-undecadienal,unii-7l26s7bw06,2,4-undecadienal, e,e,trans,trans-2,4-undecadienal,e,e-2,4-undecadien-1-al PubChem CID: 5367531 IUPAC Name: (2E,4E)-undeca-2,4-dienal SMILES: CCCCCCC=CC=CC=O