accessibility menu, dialog, popup

UserName

CAS RN 3069-40-7

H3CSiOCH3OCH3OCH3

CAS RN 3069-40-7

IUPAC Name: trimethoxy(octyl)silane
Synonyms: trimethoxycaprylylsilane
Molecular Weight (g/mol): 234.41
Molecular Formula: C11H26O3Si
InChi Key: NMEPHPOFYLLFTK-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](OC)(OC)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results