accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CBOOCH3H3CCH3CH3H3C(E/Z)

CAS RN 307531-75-5

IUPAC Name: 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pent-1-en-1-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pent-1-en-1-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 322.06
Molecular Formula: C17H32B2O4
InChi Key: MXQDNQSRLNMUOP-UHFFFAOYSA-N
SMILES: CCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results