CAS RN 30964-13-7
CAS RN 30964-13-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cynarin, MedChemExpress
MedChemExpress Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.