accessibility menu, dialog, popup

UserName

CH3OOOHO

CAS RN 3153-44-4

IUPAC Name: 4-(4-methoxyphenyl)-4-oxobutanoic acid
Synonyms: 4-(4-methoxyphenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: OMTDIBZSUZNVJK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results