accessibility menu, dialog, popup

UserName

CAS RN 338454-30-1

BOCH3HOOH

CAS RN 338454-30-1

IUPAC Name: [4-(benzyloxy)-3-methylphenyl]boronic acid
Synonyms: 4-(benzyloxy)-3-methylphenylboronic acid
Molecular Weight (g/mol): 242.08
Molecular Formula: C14H15BO3
InChi Key: QPXFEWQLALNXEZ-UHFFFAOYSA-N
SMILES: CC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results