missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methylbenzeneboronic acid, 96%

Product Code. 15439847
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15439847

Brand: Thermo Scientific Alfa Aesar H53073.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 338454-30-1 | C14H15BO3 | 242.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338454-30-1
Molecular Formula C14H15BO3
Molecular Weight (g/mol) 242.081
MDL Number MFCD06409944
InChI Key QPXFEWQLALNXEZ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid
PubChem CID 17750522
IUPAC Name (3-methyl-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O

Specifications

Melting Point 102°C to 106°C
Quantity 5 g
Formula Weight 242.08
Percent Purity 96%
Chemical Name or Material 4-Benzyloxy-3-methylbenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.