CAS RN 33953-37-6
CAS RN 33953-37-6
IUPAC Name:
N-(piperidin-4-yl)benzamide
Synonyms:
N-(piperidin-4-yl)benzamide
Molecular Weight (g/mol):
204.27
Molecular Formula:
C12H16N2O
InChi Key:
JMQDNLCNCDSHNC-UHFFFAOYSA-N
SMILES:
O=C(NC1CCNCC1)C1=CC=CC=C1
1
–
2
of
2
results
1
N-(4-Piperidyl)benzamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Benzamidopiperidine hydrate, 98%, Thermo Scientific™
CAS: 33953-37-6 Molecular Formula: C12H16N2O Molecular Weight (g/mol): 204.27 MDL Number: MFCD01314226 InChI Key: JMQDNLCNCDSHNC-UHFFFAOYSA-N Synonym: n-piperidin-4-yl-benzamide,n-piperidin-4-yl benzamide,n-4-piperidyl benzamide,4-benzamidopiperidine,n-4-piperidylbenzamide,benzamide, n-4-piperidinyl,4-benzimido piperidine,n-4-piperidinyl benzamide,4-ben-zamidopiperidine PubChem CID: 118120 IUPAC Name: N-piperidin-4-ylbenzamide SMILES: O=C(NC1CCNCC1)C1=CC=CC=C1