CAS RN 34540-22-2
CAS RN 34540-22-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Madecassoside, MedChemExpress
MedChemExpress Madecassoside is a pentacyclic triterpene isolated from Centella asitica (L.