accessibility menu, dialog, popup

UserName

CH3NBrNH2

CAS RN 34552-14-2

IUPAC Name: 2-bromo-5-methylpyridin-3-amine
Synonyms: 2-bromo-5-methylpyridin-3-amine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: ODOCHVONAOTFEQ-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(Br)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results