accessibility menu, dialog, popup

UserName

CAS RN 3460-23-9

IUPAC Name: N-(4-bromo-2-chlorophenyl)acetamide
Synonyms: N-(4-bromo-2-chlorophenyl)acetamide
Molecular Weight (g/mol): 248.5
Molecular Formula: C8H7BrClNO
InChi Key: MITWNEIUIPGZKR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results