accessibility menu, dialog, popup

UserName

ONHFBr

CAS RN 346723-41-9

IUPAC Name: 4-bromo-N-(4-fluorophenyl)benzamide
Synonyms: 4-bromo-N-(4-fluorophenyl)benzamide
Molecular Weight (g/mol): 294.12
Molecular Formula: C13H9BrFNO
InChi Key: DBCKBDOQSKIIOA-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC(=O)C2=CC=C(Br)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results